Assay Details
| Gene Symbol: TIGD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270903.1 | NP_001257832.1 | 1 | 121 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001270903.1 transcripts.
Set Membership: |
|

TIGD4
Genomic Map