Assay Details
| Gene Symbol: CHPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002816.2 | XP_009001064.1 | 4 | 2627 | 109 | |
| XM_009002818.2 | XP_009001066.1 | 3 | 2444 | 109 | ||
| GenBank mRNA | EF215029.1 | - | 1 | 220 | 109 | |
More Information
Set Membership: |
|

CHPF2
Genomic Map