Assay Details
| Gene Symbol: MPZL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760421.3 | XP_002760467.1 | 3 - 4 | 665 | 75 | |
| XM_008985090.2 | XP_008983338.1 | 3 - 4 | 665 | 75 | ||
More Information
Set Membership: |
|

MPZL1
Genomic Map