Assay Details
| Gene Symbol: PET100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761668.4 | XP_002761714.1 | 3 - 4 | 218 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PET100| Gene Symbol: PET100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761668.4 | XP_002761714.1 | 3 - 4 | 218 | 74 | |
Set Membership: |
|