Assay Details
| Gene Symbol: HINFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754432.4 | XP_002754478.2 | 2 - 3 | 303 | 65 | |
| XM_009006974.2 | XP_009005222.1 | 2 - 3 | 307 | 65 | ||
| XM_017977607.1 | XP_017833096.1 | 2 - 3 | 307 | 65 | ||
More Information
Set Membership: |
|

HINFP
Genomic Map