Assay Details
| Gene Symbol: COLEC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759270.4 | XP_002759316.1 | 6 | 932 | 99 | |
| XM_017965622.1 | XP_017821111.1 | 6 | 795 | 99 | ||
More Information
Set Membership: |
|

COLEC10
Genomic Map