Assay Details
| Gene Symbol: ZDHHC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761218.3 | XP_002761264.1 | 4 - 5 | 809 | 84 | |
| XM_008986318.1 | XP_008984566.1 | 4 - 5 | 875 | 84 | ||
| XM_008986319.2 | XP_008984567.1 | 4 - 5 | 692 | 84 | ||
| XM_008986320.2 | XP_008984568.1 | 4 - 5 | 636 | 84 | ||
| XM_017966984.1 | XP_017822473.1 | 3 - 4 | 1774 | 84 | ||
| XM_017966985.1 | XP_017822474.1 | 3 - 4 | 1702 | 84 | ||
| XM_017966986.1 | XP_017822475.1 | 4 - 5 | 2387 | 84 | ||
More Information
Set Membership: |
|

ZDHHC7
Genomic Map