Assay Details
| Gene Symbol: FAM149A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992483.2 | XP_008990731.2 | 13 - 14 | 1922 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM149A| Gene Symbol: FAM149A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992483.2 | XP_008990731.2 | 13 - 14 | 1922 | 66 | |
Set Membership: |
|