Assay Details
| Gene Symbol: SLC22A18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008687.2 | XP_009006935.1 | 6 - 7 | 950 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A18| Gene Symbol: SLC22A18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008687.2 | XP_009006935.1 | 6 - 7 | 950 | 86 | |
Set Membership: |
|