Assay Details
| Gene Symbol: NDUFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745347.4 | XP_002745393.1 | 3 - 4 | 1142 | 136 | |
| XM_017970238.1 | XP_017825727.1 | 3 - 4 | 366 | 136 | ||
More Information
Set Membership: |
|

NDUFC1
Genomic Map