Assay Details
| Gene Symbol: FUT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979297.2 | XP_008977545.1 | 3 - 4 | 881 | 67 | |
| XM_008979298.2 | XP_008977546.1 | 3 - 4 | 754 | 67 | ||
| XM_008979299.2 | XP_008977547.1 | 2 - 3 | 374 | 67 | ||
More Information
Set Membership: |
|

FUT10
Genomic Map