Assay Details
| Gene Symbol: DFFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733328.3 | XP_003733376.1 | 5 - 6 | 1006 | 53 | |
| XM_009000305.2 | XP_008998553.1 | 5 - 6 | 1028 | 53 | ||
| XM_009000306.2 | XP_008998554.1 | 5 - 6 | 1258 | 53 | ||
| XM_017974253.1 | XP_017829742.1 | 4 - 5 | 1565 | 53 | ||
More Information
Set Membership: |
|

DFFB
Genomic Map