Assay Details
| Gene Symbol: SLC16A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748846.3 | XP_002748892.2 | 4 - 5 | 3906 | 60 | |
| XM_008997952.2 | XP_008996200.1 | 4 - 5 | 1201 | 60 | ||
| XM_008997953.2 | XP_008996201.1 | 4 - 5 | 1415 | 60 | ||
| XM_008997954.2 | XP_008996202.1 | 4 - 5 | 1273 | 60 | ||
| XM_008997955.2 | XP_008996203.1 | 4 - 5 | 1290 | 60 | ||
More Information
Set Membership: |
|

SLC16A3
Genomic Map