Assay Details
| Gene Symbol: FAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986718.2 | XP_008984966.1 | 5 - 6 | 620 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM3B| Gene Symbol: FAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986718.2 | XP_008984966.1 | 5 - 6 | 620 | 72 | |
Set Membership: |
|