Assay Details
| Gene Symbol: RDH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752582.3 | XP_002752628.1 | 3 - 4 | 1280 | 60 | |
| XM_009003950.2 | XP_009002198.1 | 3 - 4 | 1299 | 60 | ||
More Information
Set Membership: |
|

RDH5
Genomic Map