Assay Details
| Gene Symbol: PECR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749734.3 | XP_002749780.2 | 3 - 4 | 487 | 63 | |
| XM_017973702.1 | XP_017829191.1 | 3 - 4 | 364 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017973702.1 transcripts.
Set Membership: |
|

PECR
Genomic Map