Assay Details
| Gene Symbol: FBXO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747119.3 | XP_002747165.1 | 2 - 3 | 930 | 91 | |
| XM_008995283.2 | XP_008993531.1 | 2 - 3 | 1091 | 91 | ||
More Information
Set Membership: |
|

FBXO5
Genomic Map