Assay Details
| Gene Symbol: LGI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762008.3 | XP_002762054.1 | 8 - 9 | 3048 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LGI4| Gene Symbol: LGI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762008.3 | XP_002762054.1 | 8 - 9 | 3048 | 57 | |
Set Membership: |
|