Assay Details
| Gene Symbol: SLC25A30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749030.4 | XP_002749076.1 | 2 - 3 | 475 | 60 | |
| XM_008998223.2 | XP_008996471.1 | 2 - 3 | 254 | 60 | ||
| XM_008998224.2 | XP_008996472.1 | 3 - 4 | 204 | 60 | ||
| XM_017973092.1 | XP_017828581.1 | 2 - 3 | 254 | 60 | ||
More Information
Set Membership: |
|

SLC25A30
Genomic Map