Assay Details
| Gene Symbol: HDDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998385.2 | XP_008996633.1 | 2 - 3 | 202 | 61 | |
| XM_017973907.1 | XP_017829396.1 | 2 - 3 | 200 | 61 | ||
| XM_017973908.1 | XP_017829397.1 | 2 - 3 | 202 | 61 | ||
More Information
Set Membership: |
|

HDDC3
Genomic Map