Assay Details
| Gene Symbol: ASCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752913.3 | XP_002752959.1 | 1 | 238 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002752913.3 transcripts.
Set Membership: |
|

ASCL1
Genomic Map