Assay Details
| Gene Symbol: SLC26A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744363.3 | XP_002744409.1 | 2 - 3 | 973 | 80 | |
| XM_008987746.2 | XP_008985994.1 | 2 - 3 | 803 | 80 | ||
More Information
Set Membership: |
|

SLC26A2
Genomic Map