Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755859.3 | XP_002755905.1 | 2 - 3 | 712 | 60 | |
| XM_009009105.2 | XP_009007353.1 | 2 - 3 | 712 | 60 | ||
More Information
Set Membership: |
|

ZNF500
Genomic Map