Assay Details
| Gene Symbol: EMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752121.4 | XP_002752167.1 | 4 - 5 | 706 | 99 | |
| XM_009003300.2 | XP_009001548.1 | 4 - 5 | 496 | 99 | ||
More Information
Set Membership: |
|

EMP1
Genomic Map