Assay Details
| Gene Symbol: GCNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995291.2 | XP_008993539.1 | 2 | 469 | 108 | |
| XM_017972875.1 | XP_017828364.1 | 3 | 564 | 108 | ||
More Information
Set Membership: |
|

GCNT1
Genomic Map