Assay Details
| Gene Symbol: LCMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755980.4 | XP_002756026.1 | 8 - 9 | 940 | 66 | |
| XM_009009343.1 | XP_009007591.1 | 6 - 7 | 790 | 66 | ||
More Information
Set Membership: |
|

LCMT1
Genomic Map