Assay Details
| Gene Symbol: NUDT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755594.4 | XP_002755640.1 | 1 - 2 | 246 | 62 | |
| XM_009008582.2 | XP_009006830.1 | 1 - 2 | 248 | 62 | ||
| XM_009008584.2 | XP_009006832.1 | 1 - 2 | 243 | 62 | ||
More Information
Set Membership: |
|

NUDT8
Genomic Map