Assay Details
| Gene Symbol: CCT6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806847.4 | XP_002806893.2 | 10 - 11 | 1365 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCT6B| Gene Symbol: CCT6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806847.4 | XP_002806893.2 | 10 - 11 | 1365 | 72 | |
Set Membership: |
|