Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752134.4 | XP_002752180.1 | 4 | 85 | ||
| XM_009003310.2 | XP_009001558.1 | 4 | 85 | |||
| GenBank mRNA | EF215134.1 | - | 1 | 321 | 85 | |
| EF215306.1 | - | 1 | 321 | 85 | ||
More Information
Set Membership: |
|

MANSC1
Genomic Map