Assay Details
| Gene Symbol: MAP3K8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750140.3 | XP_002750186.1 | 6 - 7 | 1397 | 57 | |
| XM_009000127.2 | XP_008998375.1 | 6 - 7 | 1641 | 57 | ||
| XM_017974162.1 | XP_017829651.1 | 6 - 7 | 1404 | 57 | ||
More Information
Set Membership: |
|

MAP3K8
Genomic Map