Assay Details
| Gene Symbol: BOLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964230.1 | XP_017819719.1 | 2 - 3 | 253 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BOLA3| Gene Symbol: BOLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964230.1 | XP_017819719.1 | 2 - 3 | 253 | 70 | |
Set Membership: |
|