Assay Details
| Gene Symbol: PRELID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744380.4 | XP_002744426.2 | 6 - 7 | 606 | 71 | |
| XM_003732246.3 | XP_003732294.1 | 5 - 6 | 573 | 71 | ||
| XM_008987043.2 | XP_008985291.1 | 6 - 7 | 562 | 71 | ||
| XM_017968611.1 | XP_017824100.1 | 6 - 7 | 608 | 71 | ||
| XM_017968613.1 | XP_017824102.1 | 6 - 7 | 608 | 71 | ||
| XM_017968616.1 | XP_017824105.1 | 6 - 7 | 608 | 71 | ||
| XM_017968618.1 | XP_017824107.1 | 6 - 7 | 608 | 71 | ||
| XM_017968624.1 | XP_017824113.1 | 6 - 7 | 609 | 71 | ||
More Information
Set Membership: |
|

PRELID2
Genomic Map