Assay Details
| Gene Symbol: CDKN2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001200.1 | XP_008999448.1 | 2 - 3 | 1340 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDKN2C| Gene Symbol: CDKN2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001200.1 | XP_008999448.1 | 2 - 3 | 1340 | 73 | |
Set Membership: |
|