Assay Details
| Gene Symbol: PHYHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976604.1 | XP_017832093.1 | 5 - 6 | 790 | 77 | |
| XM_017976605.1 | XP_017832094.1 | 4 - 5 | 1029 | 77 | ||
| XM_017976606.1 | XP_017832095.1 | 3 - 4 | 163 | 77 | ||
| XM_017976609.1 | XP_017832098.1 | 4 - 5 | 1210 | 77 | ||
| XM_017976613.1 | XP_017832102.1 | 4 - 5 | 1212 | 77 | ||
More Information
Set Membership: |
|

PHYHD1
Genomic Map