Assay Details
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733604.3 | XP_003733652.1 | 2 - 3 | 808 | 74 | |
| XM_009002509.2 | XP_009000757.1 | 2 - 3 | 808 | 74 | ||
| XM_017975370.1 | XP_017830859.1 | 2 - 3 | 807 | 74 | ||
| XM_017975371.1 | XP_017830860.1 | 2 - 3 | 627 | 74 | ||
| XM_017975372.1 | XP_017830861.1 | 2 - 3 | 1376 | 74 | ||
| XM_017975373.1 | XP_017830862.1 | 2 - 3 | 808 | 74 | ||
| XM_017975374.1 | XP_017830863.1 | 2 - 3 | 808 | 74 | ||
More Information
Set Membership: |
|

STEAP2
Genomic Map