Assay Details
| Gene Symbol: ANKRD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757386.3 | XP_002757432.1 | 3 - 4 | 380 | 63 | |
| XM_017964072.1 | XP_017819561.1 | 3 - 4 | 1201 | 63 | ||
More Information
Set Membership: |
|

ANKRD23
Genomic Map