Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986291.2 | XP_008984539.1 | 3 - 4 | 327 | 75 | |
| XM_008986295.2 | XP_008984543.1 | 2 - 3 | 251 | 75 | ||
| XM_017966920.1 | XP_017822409.1 | 3 - 4 | 291 | 75 | ||
More Information
Set Membership: |
|

OSGIN1
Genomic Map