Assay Details
| Gene Symbol: ACSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986380.2 | XP_008984628.1 | 3 - 4 | 951 | 65 | |
| XM_008986382.2 | XP_008984630.1 | 3 - 4 | 951 | 65 | ||
More Information
Set Membership: |
|

ACSF3
Genomic Map