Assay Details
| Gene Symbol: OGFRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746710.3 | XP_002746756.1 | 1 - 2 | 452 | 79 | |
| XM_008994653.2 | XP_008992901.1 | 1 - 2 | 452 | 79 | ||
More Information
Set Membership: |
|

OGFRL1
Genomic Map