Assay Details
| Gene Symbol: PRDX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755476.3 | XP_002755522.2 | 5 - 6 | 682 | 120 | |
| XM_009008406.2 | XP_009006654.1 | 4 - 5 | 549 | 120 | ||
| XM_017978232.1 | XP_017833721.1 | 5 - 6 | 591 | 120 | ||
More Information
Set Membership: |
|

PRDX5
Genomic Map