Assay Details
| Gene Symbol: OTUD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989259.2 | XP_008987507.1 | 1 - 2 | 598 | 125 | |
| XM_017968487.1 | XP_017823976.1 | 1 - 2 | 598 | 125 | ||
More Information
Set Membership: |
|

OTUD5
Genomic Map