Assay Details
| Gene Symbol: RPL13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762345.4 | XP_002762391.1 | 5 - 6 | 434 | 108 | |
| XM_008988394.2 | XP_008986642.1 | 5 - 6 | 353 | 108 | ||
More Information
Set Membership: |
|

RPL13A
Genomic Map