Assay Details
| Gene Symbol: GYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759508.3 | XP_002759554.1 | 7 | 1382 | 128 | |
| XM_002759509.3 | XP_002759555.1 | 8 | 1433 | 128 | ||
| XM_008983691.1 | XP_008981939.1 | 9 | 1487 | 128 | ||
| XM_008983692.1 | XP_008981940.1 | 8 | 1436 | 128 | ||
| XM_008983694.1 | XP_008981942.1 | 8 | 1305 | 128 | ||
More Information
Set Membership: |
|

GYG1
Genomic Map