Assay Details
| Gene Symbol: ZNF841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270937.1 | NP_001257866.1 | 4 - 5 | 417 | 75 | |
| XM_008988515.2 | XP_008986763.1 | 4 - 5 | 538 | 75 | ||
| XM_008988516.2 | XP_008986764.1 | 4 - 5 | 588 | 75 | ||
| XM_008988517.2 | XP_008986765.1 | 4 - 5 | 585 | 75 | ||
| XM_017967236.1 | XP_017822725.1 | 4 - 5 | 535 | 75 | ||
More Information
Set Membership: |
|

ZNF841
Genomic Map