Assay Details
| Gene Symbol: LRTOMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282060.1 | NP_001268989.1 | 4 - 5 | 989 | 94 | |
| XM_009007482.2 | XP_009005730.1 | 1 - 2 | 1821 | 94 | ||
| XM_017977497.1 | XP_017832986.1 | 2 - 3 | 920 | 94 | ||
More Information
Set Membership: |
|

LRTOMT
Genomic Map