Assay Details
| Gene Symbol: LOC100398400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734448.2 | XP_003734496.1 | 8 - 9 | 948 | 102 | |
| XM_017978936.1 | XP_017834425.1 | 8 - 9 | 948 | 102 | ||
More Information
Set Membership: |
|

LOC100398400
Genomic Map