Assay Details
| Gene Symbol: RBMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989972.2 | XP_008988220.1 | 2 - 3 | 1305 | 68 | |
| XM_017968515.1 | XP_017824004.1 | 1 - 2 | 1007 | 68 | ||
| XM_017968516.1 | XP_017824005.1 | 2 - 3 | 1067 | 68 | ||
More Information
Set Membership: |
|

RBMX
Genomic Map