Assay Details
| Gene Symbol: FATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008990041.2 | XP_008988289.1 | 2 - 3 | 362 | 87 | |
| XM_008990043.2 | XP_008988291.1 | 2 - 3 | 362 | 87 | ||
More Information
Set Membership: |
|

FATE1
Genomic Map