Assay Details
| Gene Symbol: GCM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971166.1 | XP_017826655.1 | 2 - 3 | 1125 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GCM1| Gene Symbol: GCM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971166.1 | XP_017826655.1 | 2 - 3 | 1125 | 55 | |
Set Membership: |
|