Assay Details
| Gene Symbol: ANAPC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745317.4 | XP_002745363.2 | 4 - 5 | 461 | 84 | |
| XM_008992645.2 | XP_008990893.1 | 4 - 5 | 421 | 84 | ||
More Information
Set Membership: |
|

ANAPC10
Genomic Map